missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-β-D-Glucopyranoside, MP Biomedicals™

Product Code. 11456462 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456462 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11456462 Supplier MP Biomedicals Supplier No. 0215005425
  • ONLINE EXCLUSIVE PRICE
    2240.00 €
    2121.00 € / 25g
    Save 119.00 €
    5% Off
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

n-Octyl-β-D-Glucopyranoside >98%, white powder, non-ionic detergent for membrane protein.

  • Non-ionic alkyl-glucoside detergent commonly used in the solubilization of membrane proteins and phospholipids.
  • Used at a molar dependant ratios for the solubilization of phosphatidylcholine bilayers.
  • Used for protein crystallization, as well as for enhancement of oligosaccharide cleavage from glycoproteins using endoglycosidase F.

Specifications

CAS 155751-72-7, 29836-26-8
Melting Point 89°C to 100°C (Literature)
Color White
Assay Percent Range ≥98%
Molecular Formula C14H28O6
Quantity 25 g
Beilstein 84118
Synonym okadaic acid, potassium salt, okadaic acid potassium salt, okadaic acidpotassium salt
Solubility Information Soluble in water (10mg/mL-clear colorless solution).
InChI Key UXRQUXBFVICHQJ-UHFFFAOYSA-M
Optical Rotation -31 ± 3° (c=5, water)
SMILES CC1CCC2(CCCCO2)OC1C(C)CC(C3C(=C)C(C4C(O3)CCC5(O4)CCC(O5)C=CC(C)C6CC(=CC7(O6)C(CCC(O7)CC(C)(C(=O)[O-])O)O)C)O)O.[K+]
IUPAC Name potassium;2-hydroxy-3-[11-hydroxy-2-[4-[4-hydroxy-2-[1-hydroxy-3-(3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl)butyl]-3-methylidenespiro[4a,7,8,8a-tetrahydro-4H-pyrano[3,2-b]pyran-6,5'-oxolane]-2'-yl]but-3-en-2-yl]-4-methyl-1,7-dioxaspiro[5.5]undec-4-en-8-yl]
Molecular Weight (g/mol) 292.4
PubChem CID 44134868
Formula Weight 292.4
Physical Form Powder
Chemical Name or Material n-Octyl-β-D-glucopyranoside, Molecular Biology
Grade 0.98
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.