missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltriethoxysilane, 97%

Product Code. 10566641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10566641 100 mL Glass bottle 100mL
10042011 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10566641 Supplier Thermo Scientific Acros Supplier No. 338081000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2943-75-1 | C14H32O3Si | 276.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2943-75-1
Molecular Formula C14H32O3Si
Molecular Weight (g/mol) 276.49
MDL Number MFCD00039883
InChI Key MSRJTTSHWYDFIU-UHFFFAOYSA-N
Synonym n-octyltriethoxysilane, octyltriethoxysilane, triethoxy octyl silane, silane, triethoxyoctyl, dynasylan octeo, octyl triethoxy silane, prosil 9202, silquest a 137, prosil 9234, a 137 coupling agent
PubChem CID 76262
IUPAC Name triethoxy(octyl)silane
SMILES CCCCCCCC[Si](OCC)(OCC)OCC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8800g/mL
Boiling Point 98°C
Flash Point 120°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3(CH2)7Si(OCH2CH3)3
Refractive Index 1.4160 to 1.418
Quantity 100 mL
Specific Gravity 0.88
Solubility Information Solubility in water:
Formula Weight 276.49
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material n-Octyltriethoxysilane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • n-Octyltriethoxysilane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: n-Octyltriethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.