missing translation for 'onlineSavingsMsg'
Learn More

N-Oleoyl glycine, MedChemExpress

Product Code. 16484758
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16484758 Supplier Medchem Express Supplier No. HY113204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-Oleoyl glycine is a lipoamino acid, which stimulates adipogenesis associated with activation of CB1 receptor and Akt signaling pathway in 3T3-L1 adipocyte.

N-Oleoyl glycine is a lipoamino acid, which stimulates adipogenesis associated with activation of CB1 receptor and Akt signaling pathway in 3T3-L1 adipocyte.

Specifications

Chemical Name or Material N-Oleoyl glycine
CAS 2601-90-3
Color White
Purity Grade Notes Research
Molecular Formula C20H37NO3
Solubility Information DMSO : 100 mg/mL (294.54 mM; Need ultrasonic)
SMILES O=C(O)CNC(CCCCCCC/C=C\CCCCCCCC)=O
Molecular Weight (g/mol) 339.51
Quantity 10 mg
Formula Weight 339.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.