missing translation for 'onlineSavingsMsg'
Learn More

N-Palmitoyl-D-erythro-sphingosine, synthetical, 98%

Product Code. 10413045 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10413045 25 mg Glass bottle 25mg
10266101 5 mg Glass bottle 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10413045 Supplier Thermo Scientific Acros Supplier No. 331540250
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24696-26-2 | C34H67NO3 | 537.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24696-26-2
Molecular Formula C34H67NO3
Molecular Weight (g/mol) 537.91
MDL Number MFCD00056205
InChI Key YDNKGFDKKRUKPY-UHFFFAOYNA-N
Synonym c16-ceramide, d-erythro-c16-ceramide, n-palmitoyl 4-sphingenine, n-palmitoyl-d-sphingosine, d-erythro-4-ceramide, n-palmitoylsphingosinee, d-erythro-n-palmitoylsphingosine, n-hexadecanoyl-d-erythro-ceramide, d-erythro-n-hexadecanoylsphingenine, 2s,3r,4e-2-n-palmitoyloctadecasphinga-4-ene
PubChem CID 71314645
SMILES CCCCCCCCCCCCCCCC(=O)NC(CO)C(O)C=CCCCCCCCCCCCCC

Specifications

Melting Point 94.0°C to 95.0°C
Color White
Enantiomeric Excess 99.0% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 mg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in chloroform,soluble in dichloromethane,insoluble in acetone
IUPAC Name N-[(2S,3S)-1,3-dihydroxyoctadec-4-en-2-yl]hexadecanamide
Formula Weight 537.91
Percent Purity 98% (99+% E.E.)
Physical Form Crystalline Powder
Chemical Name or Material N-Palmitoyl-D-erythro-sphingosine, Synthetical, ≥99% ee
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.