missing translation for 'onlineSavingsMsg'
Learn More

N-Palmitoyl-O-phosphocholine Serine, TRC

Product Code. 30009343
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009343 5 mg 5mg
30005578 25 mg 25mg
30004251 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009343 Supplier Toronto Research Chemicals Supplier No. TRCP3479205MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2517555-53-0
Molecular Formula C24 H49 N2 O7 P
Molecular Weight (g/mol) 508.629
Synonym (S)-2-Carboxy-2-palmitamidoethyl (2-(Trimethylammonio)ethyl)phosphate
IUPAC Name [(2S)-2-carboxy-2-(hexadecanoylamino)ethyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)N[C@@H](COP(=O)([O-])OCC[N+](C)(C)C)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H49N2O7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(27)25-22(24(28)29)21-33-34(30,31)32-20-19-26(2,3)4/h22H,5-21H2,1-4H3,(H2-,25,27,28,29,30,31)/t22-/m0/s1
Formula Weight 508.328
Percent Purity >95%
Chemical Name or Material N-Palmitoyl-O-phosphocholine Serine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.