missing translation for 'onlineSavingsMsg'
Learn More

n-Pentadecanoic acid, 99%

Product Code. 10284840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10284840 25 g Glass bottle 25g
10122990 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10284840 Supplier Thermo Scientific Acros Supplier No. 129830250
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1002-84-2 | C15H30O2 | 242.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-84-2
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.4
MDL Number MFCD00002745
InChI Key WQEPLUUGTLDZJY-UHFFFAOYSA-N
Synonym pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure
PubChem CID 13849
ChEBI CHEBI:42504
IUPAC Name pentadecanoic acid
SMILES CCCCCCCCCCCCCCC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 51°C to 53°C
Color White
Boiling Point 339°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)13CO2H
Quantity 25 g
Beilstein 02, 369
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 242.4
Percent Purity 99%
Physical Form Powder or Flakes
Chemical Name or Material n-Pentadecanoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • n-Pentadecanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.