missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane, 99+%, for residue analysis, ECD tested for pesticide analysis

Product Code. 10756812 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10756812 2.5 L Glass Bottle 2.5 L
10498872 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10756812 Supplier Thermo Scientific Acros Supplier No. 326640025
  • ONLINE EXCLUSIVE PRICE
    203.00 €
    197.00 € / 2.5 L
    Save 6.00 €
    2% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 109-66-0 | C5H12 | 72.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Spezifikation

Melting Point -130.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 0.6260g/mL
Quantity 2.5 L
Flash Point −49°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.358
Residue after Evaporation 0.0005% max.
Assay Percent Range 99+%
Specific Gravity 0.626
Linear Formula CH3(CH2)3CH3
Viscosity 0.25 mPa.s (20°C)
Water 0.005% max. (Coulometric)
Beilstein 01, 130
Merck Index 15, 7228
Boiling Point 36.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Formula Weight 72.15
Percent Purity ≥99%
Grade Residue Analysis
Chemical Name or Material n-Pentane, ECD Tested, for Pesticide Analysis
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • n-Pentane
Signalwort
  • Danger
Gefahrenkategorie
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.