missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane, BAKER ANALYZED™, J.T. Baker™

Product Code. 10751742 Shop All J.T.Baker Products
Change view
Click to view available options
:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10751742 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10751742 Supplier JT Baker Supplier No. 8114.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Laboratory Reagent

Specifications

Melting Point -130°C
CAS 109-66-0
Density 0.63 g/cm³ (20°C)
Quantity 1 L
Flash Point -40°C
Packaging Glass Bottle
Residue after Evaporation max. 0.001%
Assay Percent Range min. 99%
Molecular Formula C5H12
MDL Number MFCD00009498
Water max. 0.01%
Boiling Point 35°C to 36°C (1013 hPa)
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILES CCCCC
IUPAC Name pentane
Molecular Weight (g/mol) 72.15
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.