missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentyl-d11 Chloroformate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.50g
Specifications
Specifications
| Chemical Name or Material | Pentyl Chloroformate D11 |
| CAS | 1398065-67-2 |
| Molecular Formula | C6 D11 Cl O2 |
| Quantity | 0.5 g |
| Synonym | Amyl-d11 chloroformate, Chloroformic acid amyl ester-d11, Chloroformic acid pentyl-d11 ester, n-Pentyl-d11 chloroformate, Pentyl chloroformate D11, Undecadeuteriopentyloxycarbonyl chloride, Carbonochloridic acid pentyl-d11 ester |
| InChI Formula | InChI=1 S/C6H11ClO2/c1-2-3-4-5-9-6(7)8/h2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OC(=O)Cl |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentyl carbonochloridate |
| Molecular Weight (g/mol) | 161.671 |
| Formula Weight | 161.114 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction