missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-1-napthyl Amine, TRC

Product Code. 30416277
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30416277 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416277 Supplier Toronto Research Chemicals Supplier No. TRCP3361405G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 90-30-2
Molecular Formula C16 H13 N
Quantity 5 g
Synonym 1-Naphthalenamine, N-phenyl-, 1-Naphthylamine, N-phenyl- (7CI,8CI), 1-(N-Phenylamino)naphthalene, 1-(Phenylamino)naphthalene, 1-Anilinonaphthalene, 1-Naphthylphenylamine, Amoco 32, Antigene PA, Antioxidant A, Antioxidant PAN, C.I. 44050, Irganox L 05, N-(1-Naphthyl)-N-phenylamine, N-(1-Naphthyl)aniline, N-Phenyl-1-aminonaphthalene, N-Phenyl-1-naphthalenamine, N-Phenyl-1-naphthylamine, N-Phenyl-N-(1-naphthyl)amine, N-Phenyl-α-naphthylamine, NSC 2622, Naugard PANA, Neozone A, Nocrac PA, Nonox A, Nonox AN, PANA, Phenyl-1-naphthylamine, T 531, T 531 (antioxidant), Vulkanox PAN, α-Naphthylphenylamine
InChI Formula InChI=1S/C16H13N/c1-2-9-14(10-3-1)17-16-12-6-8-13-7-4-5-11-15(13)16/h1-12,17H
SMILES N(c1ccccc1)c2cccc3ccccc23
IUPAC Name N-phenylnaphthalen-1-amine
Molecular Weight (g/mol) 219.28
Formula Weight 219.1048
Chemical Name or Material N-Phenyl-1-naphthylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.