missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-2-naphthylamine, 97%

Product Code. 11985231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985231 100 g Glass bottle 100g
11975231 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985231 Supplier Thermo Scientific Acros Supplier No. 439911000
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 135-88-6 | C16H13N | 219.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-88-6
Molecular Formula C16H13N
Molecular Weight (g/mol) 219.29
MDL Number MFCD00004052
InChI Key KEQFTVQCIQJIQW-UHFFFAOYSA-N
Synonym n-phenyl-2-naphthylamine, n-2-naphthyl aniline, neozone, aceto pbn, stabilizator ar, agerite powder, anilinonaphthalene, antioxidant 116, pbna, 2-naphthylphenylamine
PubChem CID 8679
ChEBI CHEBI:34877
IUPAC Name N-phenylnaphthalen-2-amine
SMILES N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 105°C to 108°C
Boiling Point 395°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene,acetone,acetic acid
Formula Weight 219.28
Percent Purity 97%
Chemical Name or Material N-Phenyl-2-naphthylamine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Phenyl-2-naphthylamine
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and any thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.