missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-4-biphenylamine, TRC

Product Code. 30427832
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427832 5 mg -
30420155 50 mg -
30424843 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427832 Supplier Toronto Research Chemicals Supplier No. TRCP3365635MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32228-99-2
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.33
IUPAC Name N,4-diphenylaniline
SMILES N(c1ccccc1)c2ccc(cc2)c3ccccc3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H15N/c1-3-7-15(8-4-1)16-11-13-18(14-12-16)19-17-9-5-2-6-10-17/h1-14,19H
Formula Weight 245.1204
Chemical Name or Material N-Phenyl-4-biphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.