missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylanthranilic acid, Redox Indicator, for the detection of vanadium, ≥97.0% (T), Honeywell Fluka™

Product Code. 15697070 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15697070 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15697070 Supplier Honeywell-Fluka Supplier No. 7815010G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 182°C to 185°C (literature); 182°C to 185°C
CAS 91-40-7
CAS Max % 100%
Assay Percent Range 97.0 to 103.0%
Molecular Formula C13H11NO2
Linear Formula C20H14O4
MDL Number MFCD00005913
Beilstein 1456607
Synonym 2-(Phenylamino)benzoic acid, Diphenylamine-2-carboxylic acid, DpC
InChI Key ZWJINEZUASEZBH-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2C(=O)O
IUPAC Name 2-anilinobenzoic acid
Molecular Weight (g/mol) 213.236
PubChem CID 4386
ChEBI CHEBI:34756
Formula Weight 213.23
Grade Redox Indicator
Packaging Glass Bottle
Quantity 10 g
Chemical Name or Material N-Phenylanthranilic acid, Redox Indicator, for the detection of vanadium, ≥97.0% (T)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.