missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylanthranilic Acid, TRC

Product Code. 30423430
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423430 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423430 Supplier Toronto Research Chemicals Supplier No. TRCP308550100G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Phenylanthranilic Acid
CAS 91-40-7
Molecular Formula C13H11NO2
Quantity 100 g
Synonym 2-(Phenylamino)benzoic Acid, 2-Anilinobenzoic Acid, 2-Carboxydiphenylamine, DPC, DPC (chloride channel inhibitor), Diphenylamine-2-carboxylate, Diphenylamine-2-carboxylic Acid, Fenamic Acid, N-Phenyl-2-aminobenzoic Acid, N-Phenyl-o-aminobenzoic Acid, N-Phenylanthranilic Acid, NSC 215211, NSC 4273, o-(Phenylamino)benzoic Acid, o-Anilinobenzoic Acid, o-Carboxydiphenylamine
InChI Formula InChI=1S/C13H11NO2/c15-13(16)11-8-4-5-9-12(11)14-10-6-2-1-3-7-10/h1-9,14H,(H,15,16)
SMILES OC(=O)c1ccccc1Nc2ccccc2
IUPAC Name 2-anilinobenzoic acid
Molecular Weight (g/mol) 213.23
Formula Weight 213.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.