missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylbenzamidine, 96%, BLD Pharm™

Product Code. 30776441
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30776441 1 g 1g
30775204 100 g 100g
30770762 5 g 5g
30773252 25 g 25g
30767423 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30776441 Fornitore BLD Pharm N. del fornitore BD146171g
 Restricted Item
Estimated Shipment: 25-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Specifications

CAS 1527-91-9
Molecular Formula C13H12N2
MDL Number MFCD00019730
Quantity 1 g
SMILES N=C(NC1=CC=CC=C1)C2=CC=CC=C2
Molecular Weight (g/mol) 196.25
Percent Purity 96%
Chemical Name or Material N-Phenylbenzamidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.