missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylcarbazole, 97%

Product Code. 15294624 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15294624 100 g 100g
15264624 5 g 5g
15274624 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15294624 Supplier Thermo Scientific Acros Supplier No. 130401000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1150-62-5 | C18H13N | 243.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1150-62-5
Molecular Formula C18H13N
Molecular Weight (g/mol) 243.31
MDL Number MFCD00004965
InChI Key VIJYEGDOKCKUOL-UHFFFAOYSA-N
PubChem CID 70851
IUPAC Name 9-phenylcarbazole
SMILES C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 94.0°C to 97.0°C
Assay Percent Range 96% min. (GC)
Quantity 100 g
Beilstein 20,I,164
Synonym n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd
Formula Weight 243.31
Percent Purity 97%
Chemical Name or Material N-Phenylcarbazole
compliance-icons compliance-icons
Product Identifier
  • 9-Phenylcarbazole, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.