missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyldiethanolamine, 95%, Thermo Scientific™

Product Code. 11325066 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11325066 2.5 kg Glass bottle 2.5kg
10254750 500 g Glass bottle 500g
10675372 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11325066 Supplier Thermo Scientific Acros Supplier No. 130490025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-07-0 , 62-53-3
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD00002845
InChI Key OJPDDQSCZGTACX-UHFFFAOYSA-N
Synonym n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
PubChem CID 8416
IUPAC Name 2-[N-(2-hydroxyethyl)anilino]ethanol
SMILES OCCN(CCO)C1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 55°C to 59°C
Color White to Brown
Boiling Point 270°C
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (HOCH2CH2)2NC6H5
Quantity 2.5 kg
Solubility Information Solubility in water: 45g/L (20°C). Other solubilities: freely soluble in ethanol,acetone,ethyl acetate,and ether (20°C)
Viscosity 1.27 mPa.s (200°C)
Formula Weight 181.23
Percent Purity 95%
Physical Form Crystalline Mass, Crystals or Chunks
Chemical Name or Material N-Phenyldiethanolamine, 90-95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Phenyldiethanolamine
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.