missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyldiethanolamine, TRC

Product Code. 30476478
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476478 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476478 Supplier Toronto Research Chemicals Supplier No. TRCN2345405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material N-Phenyldiethanolamine
CAS 120-07-0
Molecular Formula C10H15NO2
Quantity 5 g
Synonym 2,2'-(Phenylimino)bis[ethanol], 2,2'-(Phenylazanediyl)diethanol, 2,2'-(Phenylimino)diethanol, 2-[(2-Hydroxyethyl)(phenyl)amino]ethan-1-ol, 2-[(2-Hydroxyethyl)phenylamino]ethanol, 2-[N-(2-Hydroxyethyl)anilino]ethanol, 3-Phenyl-3-azapentane-1,5-diol, Bis(2-hydroxyethyl)aniline, Diethanolaminobenzene, Diethanolaniline, Diethanolphenylamine, N,N-Bis(2-hydroxyethyl)-N-phenylamine, N,N-Bis(2-hydroxyethyl)aniline, N,N-Bis(beta-hydroxyethyl)aniline, N,N-Di(2-hydroxyethyl)aniline, N,N-Di(beta-hydroxyethyl)aniline, N,N-Diethanolaniline, N,N-Dihydroxyethylaniline, N-Phenyl-N,N-diethanolamine, N-Phenyliminodiethanol, NSC 6327, Phenylbis(2-hydroxyethyl)amine, Phenyldiethanolamine, [Bis(2-hydroxyethyl)amino]benzene
InChI Formula InChI=1S/C10H15NO2/c12-8-6-11(7-9-13)10-4-2-1-3-5-10/h1-5,12-13H,6-9H2
SMILES OCCN(CCO)c1ccccc1
IUPAC Name 2-[N-(2-hydroxyethyl)anilino]ethanol
Molecular Weight (g/mol) 181.23
Formula Weight 181.1103
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.