missing translation for 'onlineSavingsMsg'
Learn More

N″-Phenylguanidine nitrate, ≥97%, Thermo Scientific™

Product Code. 10260653
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10260653 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10260653 Supplier Thermo Scientific Maybridge Supplier No. BTB10165DA
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18860-78-1
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Molecular Formula C7H10N4O3
MDL Number MFCD00213415
Quantity 1 g
Synonym phenylguanidine nitrate, 1-phenylguanidine nitrate, 2-phenylguanidine nitrate, n-phenylguanidine nitrate, 2-phenylguanidine; nitric acid, nitric acid; 2-phenylguanidine, nitric acid compound with n-phenylguanidine 1:1, phenyl guanidine nitrate
InChI Key DIHKZQUSGDYVOT-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N=C(N)N.[N+](=O)(O)[O-]
IUPAC Name nitric acid;2-phenylguanidine
Molecular Weight (g/mol) 198.182
PubChem CID 2775226
Formula Weight 198.18
Percent Purity 97%
Chemical Name or Material N''-phenylguanidine nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.