missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylmaleimide, 97%

Product Code. 10225090 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10225090 25 g Glass bottle 25g
10041350 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225090 Supplier Thermo Scientific Acros Supplier No. 130710250
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 941-69-5 | C10H7NO2 | 173.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 941-69-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00005502
InChI Key HIDBROSJWZYGSZ-UHFFFAOYSA-N
Synonym n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
PubChem CID 13662
IUPAC Name 1-phenylpyrrole-2,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)C=CC2=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 86°C to 90°C
Color Yellow
Boiling Point 162°C to 163°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 400
Fieser 01,845; 13,317
Merck Index 15, 7410
Formula Weight 173.17
Percent Purity 97%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material N-Phenylmaleimide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N-Phenylmaleimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.