missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylmaleimide, 98+%

Product Code. 11432058
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432058 100 g 100g
11422058 25 g 25g
11442058 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432058 Supplier Thermo Scientific Alfa Aesar Supplier No. A14616.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 941-69-5 | C10H7NO2 | 173.171 g/mol

N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.

Solubility
Soluble in water (slightly ), methanol, ethanol, and benzene.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 941-69-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00005502
InChI Key HIDBROSJWZYGSZ-UHFFFAOYSA-N
Synonym n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
PubChem CID 13662
IUPAC Name 1-phenylpyrrole-2,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)C=CC2=O

Specifications

Melting Point 87°C to 91°C
Boiling Point 162°C to 163°C (12 mmHg)
Quantity 100 g
UN Number UN2811
Beilstein 125098
Merck Index 14,7299
Solubility Information Soluble in water (slightly ),methanol,ethanol,and benzene.
Formula Weight 173.17
Percent Purity ≥98%
Chemical Name or Material N-Phenylmaleimide
compliance-icons
Product Identifier
  • N-Phenylmaleimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: N-Phenylmaleimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.