missing translation for 'onlineSavingsMsg'
Learn More

N-Piperidineethanol, 99%

Product Code. 10246100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246100 5 g Glass bottle 5g
10266100 100 g Glass bottle 100g
10498680 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10246100 Supplier Thermo Scientific Acros Supplier No. 152110050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3040-44-6 | C7H15NO | 129.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3040-44-6
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.2
MDL Number MFCD00006512
InChI Key KZTWONRVIPPDKH-UHFFFAOYSA-N
Synonym 2-piperidinoethanol, 1-piperidineethanol, 2-piperidin-1-yl ethanol, 1-2-hydroxyethyl piperidine, n-2-hydroxyethyl piperidine, n-piperidineethanol, 2-1-piperidinyl ethanol, n-hydroxyethyl piperidine, beta-piperidylethanol, beta-piperidinoethanol
PubChem CID 18232
ChEBI CHEBI:61238
IUPAC Name 2-piperidin-1-ylethanol
SMILES C1CCN(CC1)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 16.7°C
Color Brown
Density 0.9730g/mL
Boiling Point 198°C to 203°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4794 to 1.4814
Quantity 5 g
Beilstein 20, 25
Specific Gravity 0.973
Solubility Information Solubility in water: miscible
Viscosity 20.9 mPa.s (20°C)
Formula Weight 129.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Piperidineethanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Piperidineethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.