missing translation for 'onlineSavingsMsg'
Learn More

N-Pivaloyl-o-toluidine, 99%

Product Code. 10747972 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10747972 5 g 5g
10554721 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10747972 Supplier Thermo Scientific Acros Supplier No. 346730050
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61495-04-3 | C12H17NO | 191.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61495-04-3
MDL Number MFCD00075432
InChI Key CSGRQLUGMVFNON-UHFFFAOYSA-N
Synonym n-o-tolyl pivalamide, 2,2,2'-trimethylpropionanilide, 2,2-dimethyl-n-2-methylphenyl propanamide, n-o-tolylpivalamide, acmc-1b8p7, n-2-methylphenyl pivalamide, 2,2-dimethyl-n-o-tolyl-propionamide, 2,2-dimethyl-n-2-methyl phenyl propanamide, n-2-methyl phenyl-2,2-dimethylpropanamide
PubChem CID 2734005
IUPAC Name 2,2-dimethyl-N-(2-methylphenyl)propanamide
SMILES CC1=CC=CC=C1NC(=O)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 111°C to 115°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C12H17NO
Quantity 5 g
Molecular Weight (g/mol) 191.27
Formula Weight 191.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Pivaloyl-o-toluidine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Pivaloyl-o-toluidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.