missing translation for 'onlineSavingsMsg'
Learn More

N-Prop-2-ynylurea, Thermo Scientific™

Product Code. 10344382
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10344382 1 g 1g
10575655 5 g 5g
10743433 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10344382 Supplier Thermo Scientific Maybridge Supplier No. CD11100DA
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5221-62-5
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00052817
InChI Key LJPYJRMMPVFEKR-UHFFFAOYSA-N
Synonym 1-prop-2-ynyl urea, 1-prop-2-yn-1-yl urea, n-prop-2-ynylurea, propargylurea, n-propargylurea, 2-propynylurea, prop-2-yn-1-yl urea, prop-2-yn-1-ylurea, prop-2-ynyl-urea, 1-prop-2-ynylurea
PubChem CID 2777182
IUPAC Name prop-2-ynylurea
SMILES C#CCNC(=O)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula H2NC(O)NHCH2Cidentical toCH
Quantity 1 g
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material N-Prop-2-ynylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.