missing translation for 'onlineSavingsMsg'
Learn More

N-Propargyl Phenothiazine, TRC

Product Code. 30409071
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409071 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409071 Supplier Toronto Research Chemicals Supplier No. TRCP767370500MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4282-78-4
Molecular Formula C15H11NS
Quantity 500 mg
Synonym 10-Propargylphenothiazine, 10-(2-Propyn-1-yl)-10H-phenothiazine
InChI Formula InChI=1S/C15H11NS/c1-2-11-16-12-7-3-5-9-14(12)17-15-10-6-4-8-13(15)16/h1,3-10H,11H2
SMILES C#CCN1c2ccccc2Sc3ccccc13
IUPAC Name 10-prop-2-ynylphenothiazine
Molecular Weight (g/mol) 237.32
Formula Weight 237.0612
Chemical Name or Material N-Propargyl Phenothiazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.