missing translation for 'onlineSavingsMsg'
Learn More

N-(Propargyloxy)phthalimide, 98%, Thermo Scientific™

Product Code. 10675182 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10675182 1 g Glass bottle 1g
10205380 10 g Glass bottle 10g
10265420 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10675182 Supplier Thermo Scientific Acros Supplier No. 131490010
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 4616-63-1
Molecular Formula C11H7NO3
Molecular Weight (g/mol) 201.18
MDL Number MFCD00005890
InChI Key HBGZBVPXPDNXOV-UHFFFAOYSA-N
Synonym n-propargyloxy phthalimide, n-prop-2-ynyloxy phthalimide, 2-prop-2-yn-1-yloxy isoindoline-1,3-dione, n-propargyloxyphthalimide, 2-prop-2-yn-1-yloxy isoindole-1,3-dione, 2-prop-2-ynyloxybenzo c azoline-1,3-dione, acmc-20aka8, n-2-propynoxyphthalimide, n-2-propynyloxy phthalimide, phthalimide, n-2-propynyloxy
PubChem CID 78357
IUPAC Name 2-prop-2-ynoxyisoindole-1,3-dione
SMILES C#CCON1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 149°C to 152°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 201.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-(Propargyloxy)phthalimide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.