missing translation for 'onlineSavingsMsg'
Learn More

N-Propargylphthalimide, 98%

Product Code. 15400238
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15400238

Brand: Thermo Scientific Alfa Aesar H53429.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7223-50-9 | C11H7NO2 | 185.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7223-50-9
Molecular Formula C11H7NO2
Molecular Weight (g/mol) 185.18
MDL Number MFCD00065028
InChI Key PAZCLCHJOWLTGA-UHFFFAOYSA-N
Synonym n-propargylphthalimide, 2-prop-2-yn-1-yl isoindoline-1,3-dione, n-prop-2-ynylphthalimide, 2-2-propynyl-1h-isoindole-1,3 2h-dione, 2-prop-2-ynyl-isoindole-1,3-dione, n-propargyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-2-propynyl, 2-prop-2-ynyl isoindoline-1,3-dione, 2-prop-2-yn-1-yl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-2-propyn-1-yl
PubChem CID 81644
IUPAC Name 2-prop-2-ynylisoindole-1,3-dione
SMILES O=C1N(CC#C)C(=O)C2=CC=CC=C12

Specifications

Melting Point 148°C to 152°C
Quantity 5 g
Formula Weight 185.18
Percent Purity 98%
Chemical Name or Material N-Propargylphthalimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.