missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propyl-3,3,3-d3 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Propanol-3,3,3-D3 |
| CAS | 61844-01-7 |
| Molecular Formula | C3 2H3 H5 O |
| Quantity | 0.5 g |
| Synonym | 1-Propan-3,3,3-d3-ol (9 CI), Propyl-3,3,3-d3 alcohol (6 CI), 3,3,3-Trideuteriopropan-1-ol, 1-Propanol-3,3,3-D3 |
| InChI Formula | InChI=1 S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCO |
| IUPAC Name | 3,3,3-trideuteriopropan-1-ol |
| Molecular Weight (g/mol) | 63.114 |
| Formula Weight | 63.076 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction