missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl 3,4,5-trihydroxybenzoate, 98%

Product Code. 11438720
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438720 5 g 5g
11484223 100 g 100g
11494223 500 g 500g
11404233 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11438720 Supplier Thermo Scientific Alfa Aesar Supplier No. A10877.06
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-79-9 | C10H12O5 | 212.201 g/mol

n-Propyl 3,4,5-trihydroxybenzoate is an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein. Also used as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Propyl 3,4,5-trihydroxybenzoate is an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein. Also used as an antioxidant.

Solubility
Soluble in ethanol (50 mg/ml) and water (0.35 g/100 ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-79-9
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00002196
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N
Synonym propyl gallate, n-propyl gallate, progallin p, tenox pg, nipagallin p, gallic acid, propyl ester, gallic acid propyl ester, nipa 49, benzoic acid, 3,4,5-trihydroxy-, propyl ester, 3,4,5-trihydroxybenzene-1-propylcarboxylate
PubChem CID 4947
ChEBI CHEBI:10607
IUPAC Name propyl 3,4,5-trihydroxybenzoate
SMILES CCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 146°C to 150°C
Quantity 5 g
Beilstein 1877976
Merck Index 147859
Solubility Information Soluble in ethanol (50mg/ml) and water (0.35g/100 ml at 25°C).
Formula Weight 212.2
Percent Purity 0.98
Chemical Name or Material n-Propyl 3,4,5-trihydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.