missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl acrylate, 95%, stab. with 0.1% 4-methoxyphenol

Product Code. 11392227
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392227 10 g 10g
11302237 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392227 Supplier Thermo Scientific Alfa Aesar Supplier No. L09714.09
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 925-60-0 | C6H10O2 | 114.14 g/mol

n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 925-60-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00048693
InChI Key PNXMTCDJUBJHQJ-UHFFFAOYSA-N
Synonym n-propyl acrylate, propyl acrylate, 2-propenoic acid, propyl ester, propyl 2-propenoate, acrylic acid, propyl ester, propylacrylate, ccris 8584, 1-propyl acrylate, acrylic acid propyl, acrylic acid propyl ester
PubChem CID 13550
IUPAC Name propyl prop-2-enoate
SMILES CCCOC(=O)C=C

Specifications

Density 0.92
Boiling Point 43°C to 44°C (40 mmHg)
Flash Point 24°C (75°F)
Refractive Index 1.4123
Quantity 10 g
UN Number UN3272
Beilstein 1744954
Solubility Information Immiscible with water.
Formula Weight 114.15
Percent Purity 95%
Chemical Name or Material n-Propyl acrylate, stabilized with 0.1% 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.