missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl Gallate, MP Biomedicals™

Product Code. 11466112 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466112 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11466112 Supplier MP Biomedicals Supplier No. 0210274780
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Off-White Powder

  • Anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein
  • An ester formed by the condensation of gallic acid and propanol
  • This antioxidant has been added to foods containing oils and fats to prevent oxidation
  • An antioxidant that exhibits antimicrobial activity

Specifications

CAS 121-79-9
Melting Point 140°C to 150°C
Color Off-white
Molecular Formula C10H12O5
MDL Number MFCD00002196
Beilstein 1877976
Synonym propyl gallate, n-propyl gallate, progallin p, tenox pg, nipagallin p, gallic acid, propyl ester, gallic acid propyl ester, nipa 49, benzoic acid, 3,4,5-trihydroxy-, propyl ester, 3,4,5-trihydroxybenzene-1-propylcarboxylate
Solubility Information Soluble in ethanol (50mg/mL).
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N
SMILES CCCOC(=O)C1=CC(=C(C(=C1)O)O)O
IUPAC Name propyl 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol) 212.2
PubChem CID 4947
ChEBI CHEBI:10607
Formula Weight 212.2
Percent Purity 99.2%
Quantity 100 g
Physical Form Powder
Chemical Name or Material n-Propyl gallate, Molecular Biology
Grade 0.992
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4,5-trihydroxybenzoate de propyle
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1B
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.