missing translation for 'onlineSavingsMsg'
Learn More

n-Propylboronic acid, 95%, Thermo Scientific™

Product Code. 10456062 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
Product Code.
10456062 1g; Glass bottle
1 options
This item is not returnable. View return policy

Product Code. 10456062

Brand: Thermo Scientific Acros 370870010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 17745-45-8
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C3H9BO2
Linear Formula CH3(CH2)2B(OH)2
MDL Number MFCD01074564
Quantity 1 g
Synonym n-propylboronic acid, 1-propaneboronic acid, propaneboronic acid, boronic acid, propyl, 1-propylboronic acid, 1-propaneboronic, n-propylboronc acd, propyl boronic acid, pubchem6019
InChI Key JAQOMSTTXPGKTN-UHFFFAOYSA-N
SMILES CCCB(O)O
IUPAC Name propylboronic acid
Molecular Weight (g/mol) 87.91
PubChem CID 351065
Formula Weight 87.91
Percent Purity 95%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material n-Propylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • n-Propylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.