missing translation for 'onlineSavingsMsg'
Learn More

n-Propyltriethoxysilane, 97%

Product Code. 11460077
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460077 100 g 100g
11450077 25 g 25g
11470077 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460077 Supplier Thermo Scientific Alfa Aesar Supplier No. B20997.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2550-02-9 | C9H22O3Si | 206.36 g/mol

It is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Store away from air and oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-02-9
MDL Number MFCD00026756

Specifications

Boiling Point 179°C to 180°C
Quantity 100 g
Molecular Formula C9H22O3Si
UN Number UN1993
Beilstein 1748158
Flash Point 57°C (134°F)
Synonym propyltriethoxysilane, triethoxy propyl silane, n-propyltriethoxysilane, silane, triethoxypropyl, unii-f17otx9fg5, f17otx9fg5, dynasylan pteo, triethoxypropyl-silan, triethoxypropyl-silane, triethoxy-n-propylsilane
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol,aromatic and aliphatic hydrocarbons.
InChI Key NBXZNTLFQLUFES-UHFFFAOYSA-N
SMILES CCC[Si](OCC)(OCC)OCC
IUPAC Name triethoxy(propyl)silane
Molecular Weight (g/mol) 206.36
PubChem CID 75691
Formula Weight 206.36
Percent Purity 97%
Refractive Index 1.3965
Sensitivity Moisture Sensitive
Density 0.8916
Chemical Name or Material n-Propyltriethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Propyltriethoxysilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : n-Propyltriethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.