missing translation for 'onlineSavingsMsg'
Learn More

n-Propyltriphenylphosphonium Bromide, TRC

Product Code. 30411794
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411794 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411794 Supplier Toronto Research Chemicals Supplier No. TRCP83839050G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Propyltriphenylphosphonium Bromide
CAS 6228-47-3
Molecular Formula C21H22BrP
Quantity 50 g
Synonym Triphenylpropylphosphonium Bromide, Bromo(propyl)triphenylphosphorane, NSC 50539, Propyltriphenylphosphonium Bromide
InChI Formula InChI=1S/C21H22P.BrH/c1-2-18-22(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-17H,2,18H2,1H3;1H/q+1;/p-1
SMILES [Br-].CCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name triphenyl(propyl)phosphanium;bromide
Molecular Weight (g/mol) 385.28
Formula Weight 384.0642
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.