missing translation for 'onlineSavingsMsg'
Learn More

n-Propylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15485315
Change view
Click to view available options
Quantity:
50 mL
missing translation for 'unitSize':
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
15485315 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15485315 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H58119.AD
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 156567-57-6 | C3H7BrZn | 188.373 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 156567-57-6
Density 0.947
Molecular Formula C3H7BrZn
Linear Formula CH3CH2CH2ZnBr
MDL Number MFCD00671996
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym bromozinc 1+ ;propane, propylzinc bromide, 0.50 m in thf, propylzinc bromide solution, 0.5 m in thf, n-propylzinc bromide 0.5 m in tetrahydrofuran, n-propylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key BXIZKCIGQKZYGR-UHFFFAOYSA-M
SMILES CC[CH2-].[Zn+]Br
IUPAC Name bromozinc(1+);propane
Molecular Weight (g/mol) 188.373
PubChem CID 5194901
Formula Weight 188.37
Chemical Name or Material n-Propylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.