missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 3-(propargyloxy)propionate

Product Code. 15436429
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436429 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436429 Supplier Thermo Scientific Alfa Aesar Supplier No. J64496.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1174157-65-3 | C10H11NO5 | 225.20 g/mol

N-Succinimidyl 3-(propargyloxy)propionate is an acetylene-containing reagent with NHS ester functionality for reaction with amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 3-(propargyloxy)propionate is an acetylene-containing reagent with NHS ester functionality for reaction with amines.

Solubility
Slightly soluble in water.

Notes
Keep container tightly
TRUSTED_SUSTAINABILITY

Specifications

CAS 1174157-65-3
Color Gray
Molecular Formula C10H11NO5
MDL Number MFCD13184973
Quantity 100 mg
Synonym n-succinimidyl 3-propargyloxy propionate, propargyl-nhs, ambotzpeg1935, acetylene-peg-nhs ester, propargyl-peg1-nhs ester, propargyl-succinimidyl-ester, propargyl-n-hydroxysuccinimidyl ester, 2,5-dioxopyrrolidin-1-yl 3-prop-2-ynyloxy propanoate, 2,5-dioxopyrrolidin-1-yl 3-prop-2-yn-1-yloxy propanoate
Solubility Information Slightly soluble in water.
InChI Key WKIKHHMUNOVQLD-UHFFFAOYSA-N
SMILES O=C(CCOCC#C)ON1C(=O)CCC1=O
IUPAC Name 2,5-dioxopyrrolidin-1-yl 3-(prop-2-yn-1-yloxy)propanoate
Molecular Weight (g/mol) 225.20
PubChem CID 51340986
Formula Weight 225.2
Physical Form Solid
Chemical Name or Material N-Succinimidyl 3-(propargyloxy)propionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.