missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate

Product Code. 15490397
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15490397

Brand: Thermo Scientific Alfa Aesar J64902.MB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C18H27NO9

N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate is a labeling reagent with amine reactivity as the activated NHS ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate is a labeling reagent with amine reactivity as the activated NHS ester.

Solubility
Slightly soluble in dimethyl sulfoxide and dimethyl formamide.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C18H27NO9
MDL Number MFCD22380743
Synonym Acetylene-PEG5-NHS ester

Specifications

Color Orange to Yellow
Quantity 25 mg
Solubility Information Slightly soluble in dimethyl sulfoxide and dimethyl formamide.
Formula Weight 401.41
Physical Form Viscous Liquid
Chemical Name or Material N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.