missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl Palmitate, TRC

Product Code. 30422441
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422441 1 g -
30411775 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422441 Supplier Toronto Research Chemicals Supplier No. TRCS6914001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14464-31-4
Molecular Formula C20 H35 N O4
Molecular Weight (g/mol) 353.5
IUPAC Name (2,5-dioxopyrrolidin-1-yl) hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H35NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-20(24)25-21-18(22)16-17-19(21)23/h2-17H2,1H3
Formula Weight 353.2566
Chemical Name or Material N-Succinimidyl Palmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.