missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)ethanolamine, 98%

Product Code. 10078801 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10078801

Brand: Thermo Scientific Acros 303170050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26690-80-2 | C7H15NO3 | 161.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26690-80-2
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.2
MDL Number MFCD00056657
InChI Key GPTXCAZYUMDUMN-UHFFFAOYSA-N
Synonym n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
PubChem CID 2733206
IUPAC Name tert-butyl N-(2-hydroxyethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula HOCH2CH2NHCO2C(CH3)3
Refractive Index 1.4485 to 1.4505
Density 1.0420g/mL
Quantity 5 mL
Flash Point >110°C
Formula Weight 161.2
Percent Purity 98%
Specific Gravity 1.042
Physical Form Viscous Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)ethanolamine, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.