missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butyl)thiourea, 97%, Thermo Scientific Chemicals

Product Code. 11368535
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368535 1 g 1g
11378535 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368535 Supplier Thermo Scientific Alfa Aesar Supplier No. L11979.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7204-48-0
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00041192
InChI Key RYOCWONLFFPYMN-UHFFFAOYSA-N
Synonym 1-tert-butyl-2-thiourea, 1-tert-butylthiourea, n-tert-butyl thiourea, tert-butyl-thiourea, 1-tert-butyl thiourea, t-butylthiourea, n-tertbutylthiourea, tert-butyl thiourea, n-t-butyl thiourea, acmc-1bb93
PubChem CID 737374
IUPAC Name tert-butylthiourea
SMILES CC(C)(C)NC(N)=S

Specifications

Melting Point 172°C to 176°C
Odor Odorless
Quantity 1 g
Beilstein 1744506
Formula Weight 132.23
Percent Purity 97%
Chemical Name or Material N-(tert-Butyl)thiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.