missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-tert-Butylbenzamide, 98+%

Product Code. 11338205
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338205 5 g 5g
11348205 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338205 Supplier Alfa Aesar™ Supplier No. L08509.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5894-65-5
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00041337
InChI Key HYWWTHOGCJXTRI-UHFFFAOYSA-N
Synonym n-tert-butyl benzamide, n-t-butylbenzamide, benzamide, n-tert-butyl, benzamide, n-1,1-dimethylethyl, n-t-butyl benzamide, acmc-209m8h, benzamide,1-dimethylethyl
PubChem CID 138622
IUPAC Name N-tert-butylbenzamide
SMILES CC(C)(C)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 133°C to 135°C
Quantity 5 g
Beilstein 2042391
Formula Weight 177.25
Percent Purity ≥98%
Chemical Name or Material N-tert-Butylbenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.