missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (Technical Grade), TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | MTBSTFA (with 1% t-BDMCS) |
| CAS | 77377-52-7 |
| Molecular Formula | C9 H18 F3 N O Si |
| Quantity | 100 mg |
| Synonym | MTBSTFA, N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide |
| InChI Formula | InChI=1S/C9H18F3NOSi/c1-8(2,3)15(5,6)13(4)7(14)9(10,11)12/h1-6H3 |
| SMILES | CN(C(=O)C(F)(F)F)[Si](C)(C)C(C)(C)C |
| IUPAC Name | N-[tert-butyl(dimethyl)silyl]-2,2,2-trifluoro-N-methylacetamide |
| Molecular Weight (g/mol) | 241.33 |
| Formula Weight | 241.111 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction