missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecane, 99+%

Product Code. 11367358
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367358 500 g 500g
11347358 25 g 25g
11357358 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11367358 Supplier Thermo Scientific Alfa Aesar Supplier No. 030809.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-59-4 | C14H30 | 198.394 g/mol

n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-59-4
Molecular Formula C14H30
Molecular Weight (g/mol) 198.394
MDL Number MFCD00008986
InChI Key BGHCVCJVXZWKCC-UHFFFAOYSA-N
Synonym n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
PubChem CID 12389
ChEBI CHEBI:41253
IUPAC Name tetradecane
SMILES CCCCCCCCCCCCCC

Specifications

Melting Point 6°C
Density 0.763 g/mL
Boiling Point 252°C to 254°C
Flash Point 99°C (211°F)
Assay Percent Range >99%
Refractive Index 1.429
Quantity 500 g
Solubility Information Soluble in ether,acetone and alcohol. Insoluble in water.
Formula Weight 198.39
Physical Form Liquid
Chemical Name or Material n-Tetradecane
Show More Show Less
compliance-icons
Product Identifier
  • n-Tetradecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Tetradecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.