missing translation for 'onlineSavingsMsg'
Learn More

n-Tridecane, 99+%

Product Code. 10356590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356590 25 mL Glass bottle 25mL
10531771 100 mL Glass bottle 100mL
10043903 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10356590 Supplier Thermo Scientific Acros Supplier No. 139510250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-50-5 | C13H28 | 184.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-50-5
Molecular Formula C13H28
Molecular Weight (g/mol) 184.37
MDL Number MFCD00008979
InChI Key IIYFAKIEWZDVMP-UHFFFAOYSA-N
Synonym n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group
PubChem CID 12388
ChEBI CHEBI:35998
IUPAC Name tridecane
SMILES CCCCCCCCCCCCC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -5°C to 4°C
Color Colorless
Density 0.7500g/mL
Boiling Point 234.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11CH3
Refractive Index 1.4240 to 1.4260
Quantity 25 mL
Beilstein 01, 171
Specific Gravity 0.75
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol
Formula Weight 184.36
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Tridecane
Show More Show Less
compliance-icons
Product Identifier
  • n-Tridecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.