missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)diethylamine, 97%

Product Code. 11493657
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493657 25 g 25g
11403667 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493657 Supplier Thermo Scientific Alfa Aesar Supplier No. A18149.14
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 996-50-9 | C7H19NSi | 145.32 g/mol

It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 996-50-9
Molecular Formula C7H19NSi
Molecular Weight (g/mol) 145.32
MDL Number MFCD00009040
InChI Key JOOMLFKONHCLCJ-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine
PubChem CID 70454
ChEBI CHEBI:85070
IUPAC Name N-ethyl-N-trimethylsilylethanamine
SMILES CCN(CC)[Si](C)(C)C

Specifications

Boiling Point 125°C to 126°C
Melting Point -10°C
Quantity 25 g
Flash Point 10°C (50°F)
Odor Amine-like
Refractive Index 1.411
Sensitivity Moisture sensitive
Density 0.76
UN Number UN2733
Beilstein 635718
Solubility Information Hydrolyzes in water.
Formula Weight 145.32
Percent Purity 98%
Chemical Name or Material N-(Trimethylsilyl)diethylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-(Trimethylsilyl)diethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N-(Trimethylsilyl)diethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.