missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)imidazole, 97%

Product Code. 10677522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10677522 100 g Glass bottle 100g
10317540 25 g Glass bottle 25g
10475555 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10677522 Supplier Thermo Scientific Acros Supplier No. 167131000
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

N-(Trimethylsilyl)imidazole, 97%, Quantity: 250g, Packaging: Glass bottle, Boiling Point: 93 deg.C to 94 deg.C (13.0mmHg), Colorless to Yellow, Melting Point: -42 deg.C, Molecular Weight: 140.26, Percent Purity: 97%, Assay Percent Range: 96% min. (GC), CAS: 18156-74-6 | CAS: 18156-74-6 | C6H12N2Si | 140.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
Synonym n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
PubChem CID 28925
ChEBI CHEBI:85063
IUPAC Name imidazol-1-yl(trimethyl)silane
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -42°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 93°C to 94°C (13.0 mmHg)
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4750 to 1.478
Quantity 100 g
Fieser 07,399; 11,575; 01,337; 02,177
Specific Gravity 0.95
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in most common organic solvents
Viscosity 2.5 mm2/s (25°C)
Formula Weight 140.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-(Trimethylsilyl)imidazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-(Trimethylsilyl)imidazole
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.