missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Undecane, Pure, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Packaging:
Amber glass bottle
Unit Size:
100mL
Specifications
Specifications
| CAS | 1120-21-4 |
| Melting Point | -26°C |
| Color | Colorless |
| Boiling Point | 196°C |
| Packaging | Amber glass bottle |
| Molecular Formula | C11H24 |
| MDL Number | MFCD00008959 |
| Quantity | 100 mL |
| Synonym | n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und |
| InChI Key | RSJKGSCJYJTIGS-UHFFFAOYSA-N |
| Show More |
Safety and Handling
Product Identifier
- n-Undecane
- Danger
- Aspiration toxicity Category 1
- H304-May be fatal if swallowed and enters airways.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction