missing translation for 'onlineSavingsMsg'
Learn More

N-Vinyl-epsilon-caprolactam, 99%

Product Code. 11489217
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489217 10 g 10g
11499217 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489217 Supplier Thermo Scientific Alfa Aesar Supplier No. B21311.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2235-00-9 | C8H13NO | 139.198 g/mol

N-Vinyl-epsilon-caprolactam is used as reacive diluent in a number of our screen and is used in a wide range of UV curing screen printing inks. N-Vinyl-epsilon-caprolactam is used in UV-coatings and -adhesives as well as a building block for synthesis of paper coatings. Reactive diluent for in optical fiber coatings and rapid prototyping.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Vinyl-epsilon-caprolactam is used as reacive diluent in a number of our screen and is used in a wide range of UV curing screen printing inks. N-Vinyl-epsilon-caprolactam is used in UV-coatings and -adhesives as well as a building block for synthesis of paper coatings. Reactive diluent for in optical fiber coatings and rapid prototyping.

Solubility
Soluble in water (partly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2235-00-9
Molecular Formula C8H13NO
Molecular Weight (g/mol) 139.198
MDL Number MFCD00080693
InChI Key JWYVGKFDLWWQJX-UHFFFAOYSA-N
Synonym n-vinylcaprolactam, 1-vinylazepan-2-one, n-vinyl-epsilon-caprolactam, 2h-azepin-2-one, 1-ethenylhexahydro, unii-kfc10cy9up, 1-vinylhexahydro-2h-azepin-2-one, 1-ethenylhexahydro-2h-azepin-2-one, kfc10cy9up, 1-vinylazaperhydroepin-2-one, poly-n-vinylcaprolactam
PubChem CID 75227
IUPAC Name 1-ethenylazepan-2-one
SMILES C=CN1CCCCCC1=O

Specifications

Melting Point 32°C to 36°C
Density 1.029
Boiling Point 128°C (21 mmHg)
Flash Point 101°C (214°F)
Quantity 10 g
Solubility Information Soluble in water (partly).
Formula Weight 139.2
Percent Purity 99%
Chemical Name or Material N-Vinyl-epsilon-caprolactam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.