missing translation for 'onlineSavingsMsg'
Learn More

N-Vinylphthalimide, 99%

Product Code. 11367356
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367356 1 g 1g
11377356 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367356 Supplier Thermo Scientific Alfa Aesar Supplier No. L09335.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3485-84-5 | C10H7NO2 | 173.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3485-84-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00078446
InChI Key IGDLZDCWMRPMGL-UHFFFAOYSA-N
Synonym n-vinylphthalimide, 2-vinylisoindoline-1,3-dione, phthalimide, n-vinyl, 1h-isoindole-1,3 2h-dione, 2-ethenyl, 2-vinyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1,3-dihydro-1,3-dioxoisoindole, 2-ethenyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-vinylbenzo c azolidine-1,3-dione, n-vinylphtalimide
PubChem CID 77035
IUPAC Name 2-ethenylisoindole-1,3-dione
SMILES C=CN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 84°C to 88°C
Odor Odorless
Quantity 1 g
Beilstein 127880
Formula Weight 173.17
Percent Purity 99%
Chemical Name or Material N-Vinylphthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.