missing translation for 'onlineSavingsMsg'
Learn More

N-Vinylphthalimide, 99%

Product Code. 10181002 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10181002 5 g Glass bottle 5g
10349143 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10181002 Supplier Thermo Scientific Acros Supplier No. 370200050
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3485-84-5 | C10H7NO2 | 173.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3485-84-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00078446
InChI Key IGDLZDCWMRPMGL-UHFFFAOYSA-N
Synonym n-vinylphthalimide, 2-vinylisoindoline-1,3-dione, phthalimide, n-vinyl, 1h-isoindole-1,3 2h-dione, 2-ethenyl, 2-vinyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1,3-dihydro-1,3-dioxoisoindole, 2-ethenyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-vinylbenzo c azolidine-1,3-dione, n-vinylphtalimide
PubChem CID 77035
SMILES C=CN1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 84.0°C to 88.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
IUPAC Name 2-ethenyl-2,3-dihydro-1H-isoindole-1,3-dione
Formula Weight 173.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Vinylphthalimide
Show More Show Less
compliance-icons
Product Identifier
  • N-Vinylphthalimide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.